Learn More
Kd Medical Inc PBS, 10X, PH 7.4, 4 LITER
SDP
Catalog No. 501031328
Encompass_Preferred
10X buffer solution ready for dilution to a 1X PBS working solution commonly used in biological research. Used as a wash buffer in several research applications. One 4 L cubitainer.
Catalog No. 50-103-1328 Supplier Kd Medical Inc Supplier No. PWS0204
Add to Cart
Add to Cart
10X buffer solution ready for dilution to a 1X PBS working solution commonly used in biological research. Used as a wash buffer in several research applications. One 4 L cubitainer.