Learn More
Kd Medical Inc PBS, 10X, PH7.4, 1 L BOTTLE
SDP
Catalog No. 501031326
Encompass_Preferred
10X buffer solution ready for dilution to a 1X PBS working solution commonly used in biological research. Used as a wash buffer in several research applications. One 1 Liter bottle.
Catalog No. 50-103-1326 Supplier Kd Medical Inc Supplier No. PWS0200
10X buffer solution ready for dilution to a 1X PBS working solution commonly used in biological research. Used as a wash buffer in several research applications. One 1 Liter bottle.