Learn More
Boston Bioproducts Inc Phosphate Buffered Saline, PBS (10X, pH 7.4) - 1L
Catalog No. NC9140736
Encompass_Preferred
Phosphate Buffered Saline (PBS) is a versatile buffer solution widely used due to its isotonic and non-toxic nature to cells. With a pH range of 7.0 to 7.4, PBS mimics physiological osmolarity and ion concentrations, making it ideal for applications including protein purification, Western blotting and immunoassays, sample dilution, and as a transport medium. Packaged as a 10X concentration, this solution can be diluted to support its application.
Catalog No. NC9140736 Supplier Boston Bioproducts Inc Supplier No. BM2201L
Phosphate Buffered Saline (PBS) is a versatile buffer solution widely used due to its isotonic and non-toxic nature to cells. With a pH range of 7.0 to 7.4, PBS mimics physiological osmolarity and ion concentrations, making it ideal for applications including protein purification, Western blotting and immunoassays, sample dilution, and as a transport medium. Packaged as a 10X concentration, this solution can be diluted to support its application.