Learn More
PBS, 10X Solution, Molecular Biology Grade, pH 7.4, Hoefer™

Description
Dilute to 1X with high purity H2O for a solution consisting of 9.5 mM phosphate, pH 7.4, 2.7 mM KCl and 137 mM NaCl
Specifications
Specifications
| For Use With (Application) | Sample Preparation; Immunodetection And Protein Purification |
| pH | 7.4 |
| Quantity | 1 L |
| Solution Type | Phosphate Buffered Saline |
Frequently Asked Questions (FAQs)
Our PBS, pH 7.2 and PBS, pH 7.4 products are tested for endotoxin and the endotoxin levels are shown on the Certificate of Analysis (CoA) for the particular lot number of the product.
We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625), which is molecular biology grade and is certified to be RNase-free. However, it is not sterile and hence is not cell culture-grade.
All our other PBS products (Cat. Nos. 70011xxx, 10010xxx, 70013xxx, 20012xxx) are filter-sterilized using a 0.1 µm filter and are cell culture grade (ready to use directly on cells). However, they are not tested for nuclease activity.
We offer PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free (Cat. Nos. AM9624 and AM9625) that is molecular biology grade and is certified to be RNase-free. Our other PBS products are made with distilled water and are filter-sterilized, but are not tested for RNase and DNase activity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.