Learn More
FUJIFILM Biosciences PBS 10X-Dulbecco's Phosphate Buffered Saline Solution - Liquid
Catalog No. NC9834508
Encompass
PBS 10X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.
Catalog No. NC9834508 Supplier FUJIFILM Biosciences Supplier No. 9242500ML
PBS 10X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.