Learn More
FUJIFILM BIOSCIENCES INC PBS 1X-Dulbecco's Phosphate Buffered Saline Solution - Liquid
Supplier:  FUJIFILM BIOSCIENCES INC 9240100ML
Encompass
PBS 1X Dulbecco's Phosphate Buffered Saline Solution can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability.
Catalog No. NC1404530