Learn More
Growcells PBS 1X 1L EA
Catalog No. 501034844
Encompass_Preferred
Ready to use, sterile 1X Phosphate-Buffered Saline solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 8g, KCl 0.2g, Na2HPO4 1.44g, KH2PO4 0.24g
Catalog No. 50-103-4844 Supplier Growcells Supplier No. MRGF6230
Ready to use, sterile 1X Phosphate-Buffered Saline solution prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 8g, KCl 0.2g, Na2HPO4 1.44g, KH2PO4 0.24g