Learn More
Kd Medical Inc PBS 1X, PH 7.4 W/SOD AZIDE,20L
SDP
Catalog No. 501031452
Encompass_Preferred
Phosphate Buffered Saline is used as a wash buffer in several lab applications. Contains sodium azide FACS washing grade, pH 7.4. One 20 liter cubitainer.
Catalog No. 50-103-1452 Supplier Kd Medical Inc Supplier No. RGJ0192
Add to Cart
Add to Cart
Phosphate Buffered Saline is used as a wash buffer in several lab applications. Contains sodium azide FACS washing grade, pH 7.4. One 20 liter cubitainer.