Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS, Phosphate Buffered Saline, 1X Powder, pH 7.4, Fisher BioReagents
GSA_VA

Catalog No. p-27629 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 L
50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP661-10 10 L
BP661-50 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP661-10
Only null left
Add to Cart
  • Provides a convenient way to make 10L of standard 1X phosphate buffer
  • Reconstitute contents of bottle in ultrapure water, such as BP2819, to yield a 1X PBS solution containing 0.137M NaCl, 0.0027M KCl, and 0.0119M phosphates
  • Tested for the absence of DNase, RNase, and protease
  • PBS is a standard buffer used in many biochemical applications to maintain pH
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Physical Form Powder
Quantity 10 L
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.