Learn More
Research Products International Corp PBS 1X SOLN 1ML TUBES 36/CS
SDP
Catalog No. NC1805879
Encompass_Preferred
Ready to use sterile 1X solution, supplied in sterile pre-filled screw cap tubes. Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. Each tube contains 1.0 ml PBS 1X Sterile Solution, packaged in a sealed white cardboard freezer box with a 6 x 6 divider. PBS is supplied in 4.5 ml screw cap tubes that are 3.5 “ high (cap on). Each case holds 36 pre-filled tubes.
Convenient 1.0 ml of PBS per tube
Sterile
36 x 1.0 ml
Catalog No. NC1805879 Supplier Research Products International Corp Supplier No. P1045036.0
Add to Cart
Add to Cart
Ready to use sterile 1X solution, supplied in sterile pre-filled screw cap tubes. Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. Each tube contains 1.0 ml PBS 1X Sterile Solution, packaged in a sealed white cardboard freezer box with a 6 x 6 divider. PBS is supplied in 4.5 ml screw cap tubes that are 3.5 “ high (cap on). Each case holds 36 pre-filled tubes.
Convenient 1.0 ml of PBS per tube
Sterile
36 x 1.0 ml