Learn More
Research Products International Corp PBS 200ml Tablets 100 Tab
SDP
Catalog No. 5000052402
Encompass_Preferred
PBS, 200ml Tablets refer to pre-weighed tablets used to prepare a specific volume of PBS (Phosphate Buffered Saline) solution. 
The buffer tablets are commonly used buffer solution in biological and biochemical research. PBS serves as a versatile buffer for various applications, including cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. It helps maintain a stable pH and osmolarity, providing optimal conditions for biological reactions and preserving the integrity of cells and biomolecules. 
The PBS, 200ml Tablets are designed to simplify the preparation process of PBS solutions. Each tablet contains pre-determined amounts of sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic. By using these tablets, researchers can save time and ensure accuracy by eliminating the need to weigh and measure individual components. 
Catalog No. 50-000-52402 Supplier Research Products International Corp Supplier No. P32082100T
PBS, 200ml Tablets refer to pre-weighed tablets used to prepare a specific volume of PBS (Phosphate Buffered Saline) solution. 
The buffer tablets are commonly used buffer solution in biological and biochemical research. PBS serves as a versatile buffer for various applications, including cell culture, immunohistochemistry, protein and nucleic acid research, and diagnostic assays. It helps maintain a stable pH and osmolarity, providing optimal conditions for biological reactions and preserving the integrity of cells and biomolecules. 
The PBS, 200ml Tablets are designed to simplify the preparation process of PBS solutions. Each tablet contains pre-determined amounts of sodium chloride, sodium phosphate dibasic, and potassium phosphate monobasic. By using these tablets, researchers can save time and ensure accuracy by eliminating the need to weigh and measure individual components. 