Learn More
Growcells PBS 20X 2L PK6
Catalog No. 501034857
Encompass_Preferred
Sterile 20X Phosphate-Buffered Saline solution concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 160g, KCl 4g, Na2HPO4 28.8g, KH2PO4 4.8g
Catalog No. 50-103-4857 Supplier Growcells Supplier No. MRGF63966
Sterile 20X Phosphate-Buffered Saline solution concentrate prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging. Recipe/L: NaCl 160g, KCl 4g, Na2HPO4 28.8g, KH2PO4 4.8g