Learn More
VWR INTERNATIONAL LLC Phosphate Buffered Saline (PBS) 20X, Ultra Pure Grade
Catalog No. NC1416653
Encompass
1X Solution Composition:135 mM NaCl, 2.7 mM KCl,11 mM Phosphate Buffer. Does not contain magnesium or calcium.
Catalog No. NC1416653 Supplier VWR INTERNATIONAL LLC Supplier No. 97062948
Add to Cart
Add to Cart
1X Solution Composition:135 mM NaCl, 2.7 mM KCl,11 mM Phosphate Buffer. Does not contain magnesium or calcium.