Learn More
Kd Medical Inc PBS 20X Solution, PH 7.4, 1L
SDP
Catalog No. 501031432
Encompass_Preferred
Phosphate Buffered Saline in a 20X concentrate for use as a wash buffer in immuno-labeling techniques. pH 7.4 One 1 Liter bottle.
Catalog No. 50-103-1432 Supplier Kd Medical Inc Supplier No. RGF6395
Add to Cart
Add to Cart
Phosphate Buffered Saline in a 20X concentrate for use as a wash buffer in immuno-labeling techniques. pH 7.4 One 1 Liter bottle.