Learn More
Scytek Laboratories ScyTek Phosphate Buffered Saline (25x), 7.6 pH, 1000ml
SDP
Catalog No. NC9129019
Encompass_Preferred
ScyTek Phosphate Buffered Saline (25x) pH of 7.6 is an optimal formulation of pH stabilizers and salts designed to effectively remove excess material from the microtiter plate wells without disrupting the ELISA binding reaction. By maintaining the proper buffering environment, unbound components can be washed away without suppressing antigen-antibody binding interactions, thereby reducing nonspecific background and increasing the specific signal. Our Wash buffers do not contain hazardous preservatives such as Azide or Mercury that may interfere with antibody-antigen binding interactions. For your convenience wash buffer is offered in a wide variety of formulations to meet the needs of your specific ELISA application. This product can be customized to meet the specific needs of your assay. Inquire about custom vialing, labeling, kit assembly and drop shipping.
Catalog No. NC9129019 Supplier Scytek Laboratories Supplier No. PBS999
ScyTek Phosphate Buffered Saline (25x) pH of 7.6 is an optimal formulation of pH stabilizers and salts designed to effectively remove excess material from the microtiter plate wells without disrupting the ELISA binding reaction. By maintaining the proper buffering environment, unbound components can be washed away without suppressing antigen-antibody binding interactions, thereby reducing nonspecific background and increasing the specific signal. Our Wash buffers do not contain hazardous preservatives such as Azide or Mercury that may interfere with antibody-antigen binding interactions. For your convenience wash buffer is offered in a wide variety of formulations to meet the needs of your specific ELISA application. This product can be customized to meet the specific needs of your assay. Inquire about custom vialing, labeling, kit assembly and drop shipping.