Learn More
LICORbio PBS Blocking Buffer and PVDF Kit (Single buffer kit)
Catalog No. NC2225460
Encompass_Preferred
The non-mammalian Intercept Blocker in these kits is in PBS (phosphate-buffered saline) containing 0.06% Kathon II. The Immobilon-FL PVDF membrane included in these kits has been tested to ensure a low-background for infrared Western blot success. This kit contains Intercept (PBS) Blocking Buffer (500 mL) and Millipore Immobilon-FL PVDF Membrane (1 roll; 0.45µm, 26.5 cm x 3.75 m)
This item is not returnable. View return policy
Catalog No. NC2225460 Supplier LICORbio Supplier No. 92632098
The non-mammalian Intercept Blocker in these kits is in PBS (phosphate-buffered saline) containing 0.06% Kathon II. The Immobilon-FL PVDF membrane included in these kits has been tested to ensure a low-background for infrared Western blot success. This kit contains Intercept (PBS) Blocking Buffer (500 mL) and Millipore Immobilon-FL PVDF Membrane (1 roll; 0.45µm, 26.5 cm x 3.75 m)