Learn More
Medchemexpress LLC PBS Buffer (10x) | 1 L
SDP
Catalog No. 5000428946
Encompass_Preferred
Phosphate-Buffered Saline (PBS) is a balanced salt solution primarily utilized for various cell culture applications, including washing cells, diluting cells, and preparing reagents. MedChemExpress's PBS Buffer (10x) is filtered, free of Mg2+, Ca2+, and phenol red, and has a pH ranging from 7.2 to 7.4.
  • Filtered
  • Free of Mg2+
  • Free of Ca2+
  • Free of phenol red
  • pH is 7.2-7.4
Catalog No. 50-004-28946 Supplier Medchemexpress LLC Supplier No. HYK3006500ML 2
Phosphate-Buffered Saline (PBS) is a balanced salt solution primarily utilized for various cell culture applications, including washing cells, diluting cells, and preparing reagents. MedChemExpress's PBS Buffer (10x) is filtered, free of Mg2+, Ca2+, and phenol red, and has a pH ranging from 7.2 to 7.4.
  • Filtered
  • Free of Mg2+
  • Free of Ca2+
  • Free of phenol red
  • pH is 7.2-7.4