Learn More
Beckman Coulter PBS BUFFER, 15/PK
Catalog No. NC1927314
Encompass_Preferred
This package contains 15 foil-wrapped packets of PBS Buffer reagent. Each package is brought up to a 500 mL volume with distilled water to yield a 0.01 M potassium phosphate, 0.15 M sodium chloride at a pH of 7.2 ± 0.2.
Catalog No. NC1927314 Supplier Beckman Coulter Supplier No. 6603369
This package contains 15 foil-wrapped packets of PBS Buffer reagent. Each package is brought up to a 500 mL volume with distilled water to yield a 0.01 M potassium phosphate, 0.15 M sodium chloride at a pH of 7.2 ± 0.2.