Learn More
Sigma Aldrich Fine Chemicals Biosciences Dulbecco's Phosphate Buffered Saline With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture | MFCD00131855 | 500ML
Catalog No. NC2061728
Encompass
Dulbecco's Phosphate Buffered Saline With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture | MFCD00131855 | 500ML
Catalog No. NC2061728 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D8662500ML
Dulbecco's Phosphate Buffered Saline With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture | MFCD00131855 | 500ML