Learn More
Surmodics Inc Phosphate Buffered Saline/Casein Block & Diluent-5X, 100mL
SDP
Catalog No. NC9537663
Encompass_Preferred
BioFX™ Phosphate Buffered Saline/Casein Block and Diluent-5X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.
Catalog No. NC9537663 Supplier Surmodics Inc Supplier No. PBSC010001
BioFX™ Phosphate Buffered Saline/Casein Block and Diluent-5X Concentrate minimizes non-specific interactions of the secondary reagents with each other and the primary solid phase binding sites. The high blocking efficiency of casein is due to its content of small molecular weight proteins. The solution is ready for dilution to specific application requirements.