Learn More
Bioworld PBS-Casein, 0.5%, 200 mL
Catalog No. 50198406
Encompass_Preferred
Supplied in PBS as 0.5% casein ready for use.pH range 7.4 (+/- 0.1)Sold as 2 x 100mL or 500mL.
Catalog No. 50-198-406 Supplier Bioworld Supplier No. 207200071
Supplied in PBS as 0.5% casein ready for use.pH range 7.4 (+/- 0.1)Sold as 2 x 100mL or 500mL.