Learn More
Medchemexpress LLC Pbs/cds Qds | 5 MG
SDP
Catalog No. 5000432015
Encompass_Preferred
PbS/CdS QDs (PbS/CdSe QD) is a fluorescent probe that can be used for in vivo fluorescence imaging in the second near-infrared window. The 2nd-NIR fluorescence imaging at ca.1300 nm using the PbS/CdS QDs results in the highest signal to background ratio with a low blurring for in vivo imaging.
  • Fluorescent probe for in vivo fluorescence imaging in the second near-infrared window
  • Highest signal to background ratio with low blurring for in vivo imaging at ca.1300 nm
Catalog No. 50-004-32015 Supplier Medchemexpress LLC Supplier No. HYD13915MG
Add to Cart
Add to Cart
PbS/CdS QDs (PbS/CdSe QD) is a fluorescent probe that can be used for in vivo fluorescence imaging in the second near-infrared window. The 2nd-NIR fluorescence imaging at ca.1300 nm using the PbS/CdS QDs results in the highest signal to background ratio with a low blurring for in vivo imaging.
  • Fluorescent probe for in vivo fluorescence imaging in the second near-infrared window
  • Highest signal to background ratio with low blurring for in vivo imaging at ca.1300 nm