Learn More
Medchemexpress LLC PbS/CdS QDs (PbS/CdSe QD) | PbS/CdS | 10 MG
SDP
Catalog No. 5000297184
Encompass_Preferred
PbS/CdS quantum dots are colloidal nanocrystals designed as a fluorescent probe for in vivo imaging in the second near-infrared (NIR-II) window. These aqueous suspensions emit near ~1300 nm, yielding high signal-to-background ratio and reduced tissue scattering for deeper, clearer fluorescence imaging. The product is supplied as a light yellow to brown liquid suspension at 10 mg/mL and requires protection from light during storage.
Catalog No. 50-002-97184 Supplier Medchemexpress LLC Supplier No. HYD139110MG
PbS/CdS quantum dots are colloidal nanocrystals designed as a fluorescent probe for in vivo imaging in the second near-infrared (NIR-II) window. These aqueous suspensions emit near ~1300 nm, yielding high signal-to-background ratio and reduced tissue scattering for deeper, clearer fluorescence imaging. The product is supplied as a light yellow to brown liquid suspension at 10 mg/mL and requires protection from light during storage.