Learn More
Enzo Life Sciences PBS (endotoxin-free) (sterile) (10ml)
SDP
Catalog No. 502009343
Encompass_Preferred
Sterile and endotoxin-free PBS for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Long Term Storage: +4°C.
Catalog No. 50-200-9343 Supplier Enzo Life Sciences Supplier No. ALX505007L010
Sterile and endotoxin-free PBS for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Long Term Storage: +4°C.