Learn More
AdipoGen PBS Endotoxin-free (sterile) (3 x 10 ml)
SDP
Catalog No. 501687223
Encompass_Preferred
PBS Endotoxin-free (sterile) (3 x 10 ml), Adipogen Life Sciences. Chemical. Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-168-7223 Supplier AdipoGen Supplier No. IAX9000013010
Add to Cart
Add to Cart
PBS Endotoxin-free (sterile) (3 x 10 ml), Adipogen Life Sciences. Chemical. Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).