Learn More
AdipoGen PBS Endotoxin-free (sterile) (100 ml)
SDP
Catalog No. 501490619
Encompass_Preferred
PBS Endotoxin-free (sterile) (100 ml), AdipoGen Life Sciences. Chemical. Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-149-0619 Supplier AdipoGen Supplier No. IAX900001L100
PBS Endotoxin-free (sterile) (100 ml), AdipoGen Life Sciences. Chemical. Endotoxin free buffer used for the dilution of TLRpure LPS and solubilisation of endotoxin-free reagents (e.g. ODNs).