Learn More
Bioworld PBS/Fish Gelatin (10X), 200 mL
Catalog No. 50198408
Encompass_Preferred
PBS/Fish Gelatin (1x) Phosphate Buffered Saline/fish gelatin 0.5% blocks, diluents, and coating solutions.Technical Information 1x: Fish Gelatin 0.5%PBS (1x)pH range: 7.4 (+/- 0.1)
Catalog No. 50-198-408 Supplier Bioworld Supplier No. 207200081
Add to Cart
Add to Cart
PBS/Fish Gelatin (1x) Phosphate Buffered Saline/fish gelatin 0.5% blocks, diluents, and coating solutions.Technical Information 1x: Fish Gelatin 0.5%PBS (1x)pH range: 7.4 (+/- 0.1)