Learn More
Apexbio Technology LLC PBS Liposomes(Synonyms: Phosphate Buffered Saline Liposomes, Empty Liposomes, Blank Liposomes), 5mL.
SDP
Catalog No. 5000061324
Encompass_Preferred
PBS Liposomes consist of phosphate-buffered saline (PBS) encapsulated within lipid bilayers serving primarily as control reagents in macrophage depletion studies involving clodronate liposomes Unlike clodronate-loaded liposomes PBS liposomes lack active agents targeting macrophages and thus do not induce macrophage apoptosis They function to discriminate between the biological effects mediated by liposomal delivery systems versus those specifically attributable to encapsulated clodronate PBS Liposomes are commonly employed in comparative experimental designs to assess specific macrophage depletion outcomes
Catalog No. 50-000-61324 Supplier Apexbio Technology LLC Supplier No. K27225
PBS Liposomes consist of phosphate-buffered saline (PBS) encapsulated within lipid bilayers serving primarily as control reagents in macrophage depletion studies involving clodronate liposomes Unlike clodronate-loaded liposomes PBS liposomes lack active agents targeting macrophages and thus do not induce macrophage apoptosis They function to discriminate between the biological effects mediated by liposomal delivery systems versus those specifically attributable to encapsulated clodronate PBS Liposomes are commonly employed in comparative experimental designs to assess specific macrophage depletion outcomes