Learn More
Aniara Diagnostica LLC PBS PDR PH 7.4 10L
Catalog No. 501008861
Encompass
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed powder mix in sealed pouch yields 10 L 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Used as biological buffer, isotonic. 1 pouch.
Catalog No. 50-100-8861 Supplier Aniara Diagnostica LLC Supplier No. A1294241
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed powder mix in sealed pouch yields 10 L 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Used as biological buffer, isotonic. 1 pouch.