Learn More
Crystalgen Inc PBS PH7.2 10X 500ML
SDP
Catalog No. NC2302954
Encompass_Preferred
PBS (Phosphate Buffered Saline) pH7.2 10X, with Ca, Mg-500mlUltra pure Phosphate buffered isotonic saline. With calcium and magnesium. Sterile filtered. 1X PBS solutions contains 137mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 2mM KH2PO4.pH: 7.2Storage : 18-26oC
Catalog No. NC2302954 Supplier Crystalgen Inc Supplier No. 22114112
PBS (Phosphate Buffered Saline) pH7.2 10X, with Ca, Mg-500mlUltra pure Phosphate buffered isotonic saline. With calcium and magnesium. Sterile filtered. 1X PBS solutions contains 137mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 2mM KH2PO4.pH: 7.2Storage : 18-26oC