Learn More
Alkali Scientific PBS [Phosphate Buffered Saline] 100 mL Tablets, 100 Tablets per Bottle
SDP
Catalog No. 5000661046
Encompass_Preferred
PBS [Phosphate Buffered Saline] 100 mL Tablets are solid tablets that dissolve easily to prepare 100 mL of phosphate-buffered saline. These tablets provide a convenient and efficient way to prepare PBS solutions, eliminating the need for weighing and measuring individual components. They are ideal for laboratory applications that require consistent and reliable buffer preparation, including cell culture, microbiological testing, and molecular biology procedures. The tablets dissolve quickly in water, ensuring that the resulting buffer maintains the correct pH and osmolarity for a wide range of applications. Each bottle contains 100 tablets, making it a cost-effective option for labs that require frequent PBS preparation.
Catalog No. 50-006-61046 Supplier Alkali Scientific Supplier No. PBA003CP
Add to Cart
Add to Cart
PBS [Phosphate Buffered Saline] 100 mL Tablets are solid tablets that dissolve easily to prepare 100 mL of phosphate-buffered saline. These tablets provide a convenient and efficient way to prepare PBS solutions, eliminating the need for weighing and measuring individual components. They are ideal for laboratory applications that require consistent and reliable buffer preparation, including cell culture, microbiological testing, and molecular biology procedures. The tablets dissolve quickly in water, ensuring that the resulting buffer maintains the correct pH and osmolarity for a wide range of applications. Each bottle contains 100 tablets, making it a cost-effective option for labs that require frequent PBS preparation.