Learn More
Alkali Scientific PBS [Phosphate Buffered Saline] 100 mL Tablets, 500 Tablets per Bottle
SDP
Catalog No. 5000660079
Encompass_Preferred
PBS [Phosphate Buffered Saline] 100 mL Tablets are designed to make buffer preparation simple and efficient. Each tablet dissolves easily in water to create 100 mL of phosphate-buffered saline. With 500 tablets per bottle, these tablets are perfect for laboratories with high-volume buffer preparation needs. The ease of use and consistency of the PBS solution make these tablets ideal for various applications, including cell culture, microbiological analysis, and molecular biology experiments. The tablets provide a reliable way to ensure that each PBS solution is accurately prepared with the correct pH and osmolarity, minimizing the potential for human error in buffer formulation.
Catalog No. 50-006-60079 Supplier Alkali Scientific Supplier No. PBA004CP
Add to Cart
Add to Cart
PBS [Phosphate Buffered Saline] 100 mL Tablets are designed to make buffer preparation simple and efficient. Each tablet dissolves easily in water to create 100 mL of phosphate-buffered saline. With 500 tablets per bottle, these tablets are perfect for laboratories with high-volume buffer preparation needs. The ease of use and consistency of the PBS solution make these tablets ideal for various applications, including cell culture, microbiological analysis, and molecular biology experiments. The tablets provide a reliable way to ensure that each PBS solution is accurately prepared with the correct pH and osmolarity, minimizing the potential for human error in buffer formulation.