Learn More
Alkali Scientific PBS [Phosphate Buffered Saline], 10X Powder Concentrate, White Granular Powder, 980g make, 10 L
SDP
Catalog No. 5000661556
Encompass_Preferred
PBS [Phosphate Buffered Saline] 10X Powder Concentrate, in a 980g pack, makes 10 liters of 10X PBS solution. This highly concentrated powder is perfect for laboratories that require large volumes of PBS for a wide range of biological and biochemical applications. The powder dissolves easily in water, providing a quick and convenient way to prepare the solution. 10X PBS is essential in many laboratory protocols, including protein assays, cell culture, and washing procedures. The larger pack size is ideal for high-throughput laboratories or research facilities that need to prepare PBS solutions in bulk for routine use in experiments and testing.
Catalog No. 50-006-61556 Supplier Alkali Scientific Supplier No. PBA008CP
PBS [Phosphate Buffered Saline] 10X Powder Concentrate, in a 980g pack, makes 10 liters of 10X PBS solution. This highly concentrated powder is perfect for laboratories that require large volumes of PBS for a wide range of biological and biochemical applications. The powder dissolves easily in water, providing a quick and convenient way to prepare the solution. 10X PBS is essential in many laboratory protocols, including protein assays, cell culture, and washing procedures. The larger pack size is ideal for high-throughput laboratories or research facilities that need to prepare PBS solutions in bulk for routine use in experiments and testing.