Learn More
Alkali Scientific PBS [Phosphate Buffered Saline] 200mL Tablets, 500 Tablets per Bottle
SDP
Catalog No. 5000660121
Encompass_Preferred
PBS [Phosphate Buffered Saline] 200mL Tablets are designed for laboratories that need to prepare larger volumes of PBS quickly and accurately. Each tablet dissolves easily in water to create 200 mL of phosphate-buffered saline, a standard buffer used in many biological and biochemical applications. The 500-tablet bottle is perfect for high-throughput labs that require a consistent and reliable buffer solution. These tablets ensure that the PBS solution maintains the correct pH and osmolarity, which is essential for applications such as cell culture, protein assays, and molecular biology experiments. The tablets are convenient, cost-effective, and reduce the chance of errors in solution preparation.
Catalog No. 50-006-60121 Supplier Alkali Scientific Supplier No. PBA006CP
PBS [Phosphate Buffered Saline] 200mL Tablets are designed for laboratories that need to prepare larger volumes of PBS quickly and accurately. Each tablet dissolves easily in water to create 200 mL of phosphate-buffered saline, a standard buffer used in many biological and biochemical applications. The 500-tablet bottle is perfect for high-throughput labs that require a consistent and reliable buffer solution. These tablets ensure that the PBS solution maintains the correct pH and osmolarity, which is essential for applications such as cell culture, protein assays, and molecular biology experiments. The tablets are convenient, cost-effective, and reduce the chance of errors in solution preparation.