Learn More
Research Products International Corp PBS [Phosphate Buffered Saline], 1X Solution, 4.5ml Pre-Filled Tubes, Sterile, 36 Tubes per Case,10ml Tube Size
SDP
Catalog No. 502114056
Encompass_Preferred
Ready to use sterile 1X solution, supplied in sterile pre-filled screw cap tubes. Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. Each tube contains 4.5 ml PBS 1X Sterile Solution, packaged in a sealed white cardboard freezer box with a 6 x 6 divider. PBS is supplied in 10 ml screw cap tubes that are 3.5 “ high (cap on). Each case holds 36 pre-filled tubes.
Convenient 4.5ml of PBS per tube
Sterile
36 x 4.5 ml
Catalog No. 50-211-4056 Supplier Research Products International Corp Supplier No. P1090036.0
Ready to use sterile 1X solution, supplied in sterile pre-filled screw cap tubes. Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. Each tube contains 4.5 ml PBS 1X Sterile Solution, packaged in a sealed white cardboard freezer box with a 6 x 6 divider. PBS is supplied in 10 ml screw cap tubes that are 3.5 “ high (cap on). Each case holds 36 pre-filled tubes.
Convenient 4.5ml of PBS per tube
Sterile
36 x 4.5 ml