Learn More
Biocare Medical, LLC PBS Plus, 10X
SDP
Catalog No. 5082972
Encompass_Preferred
For laboratory use and procedures requiring a high quality phosphate buffered saline with superior pH stability. PBS Plus (sodium azide free) buffer works well for manual and automated histopathology and immunohistochemistry (IHC) applications with the addition of a surfactant.
Catalog No. 50-829-72 Supplier Biocare Medical, LLC Supplier No. PBS940M
Add to Cart
Add to Cart
For laboratory use and procedures requiring a high quality phosphate buffered saline with superior pH stability. PBS Plus (sodium azide free) buffer works well for manual and automated histopathology and immunohistochemistry (IHC) applications with the addition of a surfactant.