Learn More
Boston Bioproducts Inc Phosphate Buffered Saline, PBS (Sterile-Filtered, RNase Free) - 1L
Catalog No. NC1526599
Encompass
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a pH range of 7.0 to 7.4, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, western blotting and immunoassays, sample dilution, and as a transport medium. This buffer is also tested for RNase enzymes, minimizing risk of nucleic acid degredation in downstream applications.
Catalog No. NC1526599 Supplier Boston Bioproducts Inc Supplier No. BM220SDR1L
Add to Cart
Add to Cart
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a pH range of 7.0 to 7.4, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, western blotting and immunoassays, sample dilution, and as a transport medium. This buffer is also tested for RNase enzymes, minimizing risk of nucleic acid degredation in downstream applications.