Learn More
Medchemexpress LLC PBS-T Powder | 100 POUCHES
SDP
Catalog No. 5000430380
Encompass_Preferred
MCE PBS-T Powder (1 L of 1x) is a formulation consisting of NaCl, KCl, Phosphate, and Tween 20. It is extensively utilized as a washing buffer in immunoassay experiments such as ELISA and Western Blot. Additionally, it can be employed for antibody dilution and the preparation of blocking solutions. The product is conveniently packaged, with 20 pouches defined as the base specification, and larger sizes offered as incremental volumes of this base. Each pouch is designed to be diluted to 1 L of 1x PBS-T buffer.
  • Widely used in ELISA and Western Blot.
  • Can be used for antibody dilution and preparation of blocking solution.
  • Consists of NaCl, KCl, Phosphate, and Tween 20.
  • Each pouch dilutes to 1 L of 1x PBS-T buffer.
  • Eliminates the need to weigh and mix individual components.
  • Convenient way to make PBS-T buffer.
  • Final 1x PBS-T buffer has a pH of 7.4 ± 0.2 @ 25°C.
Catalog No. 50-004-30380 Supplier Medchemexpress LLC Supplier No. HYK1022100POUCHES
MCE PBS-T Powder (1 L of 1x) is a formulation consisting of NaCl, KCl, Phosphate, and Tween 20. It is extensively utilized as a washing buffer in immunoassay experiments such as ELISA and Western Blot. Additionally, it can be employed for antibody dilution and the preparation of blocking solutions. The product is conveniently packaged, with 20 pouches defined as the base specification, and larger sizes offered as incremental volumes of this base. Each pouch is designed to be diluted to 1 L of 1x PBS-T buffer.
  • Widely used in ELISA and Western Blot.
  • Can be used for antibody dilution and preparation of blocking solution.
  • Consists of NaCl, KCl, Phosphate, and Tween 20.
  • Each pouch dilutes to 1 L of 1x PBS-T buffer.
  • Eliminates the need to weigh and mix individual components.
  • Convenient way to make PBS-T buffer.
  • Final 1x PBS-T buffer has a pH of 7.4 ± 0.2 @ 25°C.