Learn More
Aniara Diagnostica LLC Phosphate Buffered Saline with Tween 20, pH 7.4, 100 tablets
Catalog No. 501008813
Encompass
Medicago Phosphate Buffered Saline w/Tween (PBS-T) pH 7.4 supplied as preweighed tablets yields 1000 mL 0.14 M NaCl, 0.0027 M KCl, 0.05% Tween-20, 0.01 M phosphate buffer with pH 7.4 at 25C. Use for immunoassays. Bottle of 100 Tabelts.
Catalog No. 50-100-8813 Supplier Aniara Diagnostica LLC Supplier No. A099410100
Medicago Phosphate Buffered Saline w/Tween (PBS-T) pH 7.4 supplied as preweighed tablets yields 1000 mL 0.14 M NaCl, 0.0027 M KCl, 0.05% Tween-20, 0.01 M phosphate buffer with pH 7.4 at 25C. Use for immunoassays. Bottle of 100 Tabelts.