Learn More
AG Scientific Inc PBS Tablets 100 Tablets
Catalog No. 502268352
Encompass_Preferred
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.
Catalog No. 50-226-8352 Supplier AG Scientific Inc Supplier No. P2424100
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.