Learn More
Abcam PBS Tablets (1000 ml), 10TABLET
Catalog No. 502205521
Encompass_Preferred
MW 411.04 Da. One of the most commonly used buffers provided in a convenient, tablet form. PBS is isotonic and non-toxic to cells and has the ability to maintain their osmolarity thereby making the buffer suitable for washing procedures in cell cultures and for immunoassays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5521 Supplier Abcam Supplier No. AB28541010TABLET
MW 411.04 Da. One of the most commonly used buffers provided in a convenient, tablet form. PBS is isotonic and non-toxic to cells and has the ability to maintain their osmolarity thereby making the buffer suitable for washing procedures in cell cultures and for immunoassays.
The product is subject to the following: Abcam Restricted Use Statement