Learn More
Gold Biotechnology Inc PBS Tablets
SDP
Catalog No. 5000562128
Encompass_Preferred
PBS (phosphate buffered saline) is a common buffer solution used in the laboratory It is an isotonic and non-toxic buffer that is meant to mimic the osmolarity and ion concentrations of the human body PBS is used for immunoassays for sample dilution for protein purification and as a wash buffer in cell cultures PBS tablets are premeasured tablets for convenient preparation of 1X PBS solution at a pH of 7 3 7 5 PBS buffer contains 10mM phosphate buffer 137mM sodium chloride and 2 7mM potassium chloride
Catalog No. 50-005-62128 Supplier Gold Biotechnology Inc Supplier No. P271200
PBS (phosphate buffered saline) is a common buffer solution used in the laboratory It is an isotonic and non-toxic buffer that is meant to mimic the osmolarity and ion concentrations of the human body PBS is used for immunoassays for sample dilution for protein purification and as a wash buffer in cell cultures PBS tablets are premeasured tablets for convenient preparation of 1X PBS solution at a pH of 7 3 7 5 PBS buffer contains 10mM phosphate buffer 137mM sodium chloride and 2 7mM potassium chloride