Learn More
MilliporeSigma™ PBS Tablets, Calbiochem™,
Catalog No. 5246501EA Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Strip
Catalog No. Quantity
52-465-01EA 1 Strip
1 options
Catalog No. 52-465-01EA Supplier MilliporeSigma™ Supplier No. 5246501EA
Add to Cart
Add to Cart
Dissolving one tablet in 1 liter of deionized H2O yields 140mM NaCl, 10mM phosphate buffer, and 3mM KCl
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Strip
Grade Not Available