Learn More
Aniara Diagnostica LLC PBS TBLT PH 7.4 1000ML 100 EA
Catalog No. 501008811
Encompass
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed tablets yields 1000 mL 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.
Catalog No. 50-100-8811 Supplier Aniara Diagnostica LLC Supplier No. A099400100
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed tablets yields 1000 mL 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.