Learn More
Aniara Diagnostica LLC Phosphate Buffered Saline, pH 7.4, 100 tablets
Catalog No. 501008798
Encompass
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed tablets yields 500 mL 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.
Catalog No. 50-100-8798 Supplier Aniara Diagnostica LLC Supplier No. A098912100
Medicago Phosphate Buffered Saline (PBS) pH 7.4 supplied as preweighed tablets yields 500 mL 0.14 M NaCl, 0.0027 M KCl, 0.010 M phosphate buffer with pH 7.4 at 25C. Use as biological buffer, isotonic. Bottle of 100 tablets.