Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PBT434 mesylate | 2387898-69-1 | 99.9% | 50 MG
SDP

Catalog No. 5000233263
Encompass_Preferred

PBT434 mesylate is the methanesulfonate salt of a small-molecule inhibitor of α-synuclein aggregation that demonstrates oral activity and crosses the blood-brain barrier, supplied for preclinical research. It also exhibits iron-modulating properties and is provided as a high-purity solid with defined storage conditions.

  • High purity: 99.9%.
  • Molecular weight 398.26 g·mol⁻1.
  • Chemical formula C13H17Cl2N3O5S.
  • Available in small pack sizes including 50 MG.
  • Store sealed at 4°C; in solvent keep at -80°C up to 6 months, or -20°C up to 1 month.
  • Intended for preclinical studies of protein aggregation and iron modulation; follow appropriate safety guidance.

Catalog No. 50-002-33263 Supplier Medchemexpress LLC Supplier No. HY120475A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PBT434 mesylate is the methanesulfonate salt of a small-molecule inhibitor of α-synuclein aggregation that demonstrates oral activity and crosses the blood-brain barrier, supplied for preclinical research. It also exhibits iron-modulating properties and is provided as a high-purity solid with defined storage conditions.

  • High purity: 99.9%.
  • Molecular weight 398.26 g·mol⁻1.
  • Chemical formula C13H17Cl2N3O5S.
  • Available in small pack sizes including 50 MG.
  • Store sealed at 4°C; in solvent keep at -80°C up to 6 months, or -20°C up to 1 month.
  • Intended for preclinical studies of protein aggregation and iron modulation; follow appropriate safety guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.