Learn More
Medchemexpress LLC PBT434 methanesulfonate | 2387898-69-1 | 99.9% | 1 MG
SDP
Catalog No. 5000234340
Encompass_Preferred
PBT434 mesylate (PBT434 methanesulfonate, ATH434) is a brain-penetrant, orally active small molecule inhibitor of α-synuclein aggregation with iron-chelating activity, supplied for research use in studies of synucleinopathies and neurodegeneration.
  • Inhibits α-synuclein aggregation and reduces iron-mediated aggregation.
  • Acts as an iron chelator and modulates iron trafficking.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • High purity appropriate for biochemical and cellular assays.
  • Solid, white to off-white appearance; store sealed at 4°C.
Catalog No. 50-002-34340 Supplier Medchemexpress LLC Supplier No. HY120475A1MG
PBT434 mesylate (PBT434 methanesulfonate, ATH434) is a brain-penetrant, orally active small molecule inhibitor of α-synuclein aggregation with iron-chelating activity, supplied for research use in studies of synucleinopathies and neurodegeneration.
  • Inhibits α-synuclein aggregation and reduces iron-mediated aggregation.
  • Acts as an iron chelator and modulates iron trafficking.
  • Brain-penetrant and orally active, suitable for in vivo studies.
  • High purity appropriate for biochemical and cellular assays.
  • Solid, white to off-white appearance; store sealed at 4°C.