Learn More
Medchemexpress LLC PBT434 methanesulfonate | 2387898-69-1 | 99.9% | 10 MG
SDP
Catalog No. 5000233527
Encompass_Preferred
PBT434 mesylate is the methanesulfonate salt of PBT434, an orally active, blood-brain barrier-penetrant small molecule that inhibits α-synuclein aggregation and modulates iron-mediated redox activity for preclinical neurodegeneration research.
  • Orally active α-synuclein aggregation inhibitor.
  • Crosses the blood-brain barrier.
  • High purity suitable for research use.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic agitation recommended.
  • Store solid at 4°C sealed; stock solutions tolerate -80°C for long-term storage.
Catalog No. 50-002-33527 Supplier Medchemexpress LLC Supplier No. HY120475A10MG
PBT434 mesylate is the methanesulfonate salt of PBT434, an orally active, blood-brain barrier-penetrant small molecule that inhibits α-synuclein aggregation and modulates iron-mediated redox activity for preclinical neurodegeneration research.
  • Orally active α-synuclein aggregation inhibitor.
  • Crosses the blood-brain barrier.
  • High purity suitable for research use.
  • Soluble in DMSO at approximately 100 mg/mL; ultrasonic agitation recommended.
  • Store solid at 4°C sealed; stock solutions tolerate -80°C for long-term storage.