Learn More
Medchemexpress LLC PBX-7011 TFA | 3003834-30-5 | 98.6% | 561.46 | 1 ML
SDP
Catalog No. 5000391691
Encompass_Preferred
PBX-7011 TFA is a derivative of camptothecin that inhibits the expression of cancer-related survival genes and degrades DDX5 proteins.
  • Inhibits expression of cancer-related survival genes (DDX5, Survivin, Mcl-1, and XIAP in FaDu cells).
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.
Catalog No. 50-003-91691 Supplier Medchemexpress LLC Supplier No. HY160438B10MM/1ML
PBX-7011 TFA is a derivative of camptothecin that inhibits the expression of cancer-related survival genes and degrades DDX5 proteins.
  • Inhibits expression of cancer-related survival genes (DDX5, Survivin, Mcl-1, and XIAP in FaDu cells).
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.