Learn More
Medchemexpress LLC PBX-7011 TFA | 98.6% | 561.46 | 25 MG
SDP
Catalog No. 5000393008
Encompass_Preferred
PBX-7011 TFA is a derivative of camptothecin that exhibits anticancer activity. It functions by inhibiting the expression of cancer-related survival genes, specifically DDX5, Survivin, Mcl-1, and XIAP, in FaDu cells. Additionally, it degrades DDX5 proteins.
  • Derivative of camptothecin.
  • Inhibits expressions of cancer related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells.
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.
Catalog No. 50-003-93008 Supplier Medchemexpress LLC Supplier No. 25MGHY160438B
PBX-7011 TFA is a derivative of camptothecin that exhibits anticancer activity. It functions by inhibiting the expression of cancer-related survival genes, specifically DDX5, Survivin, Mcl-1, and XIAP, in FaDu cells. Additionally, it degrades DDX5 proteins.
  • Derivative of camptothecin.
  • Inhibits expressions of cancer related survival genes DDX5, Survivin, Mcl-1, and XIAP in FaDu cells.
  • Degrades DDX5 proteins.
  • Exhibits anticancer activity.